Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexanemethanol, 99%

Catalog No. AC111340250
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC111340250 25 g Glass bottle
AC111340050 5 g Glass bottle
AC111341000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC111340250 Supplier Thermo Scientific Chemicals Supplier No. 111340250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 100-49-2 | C7H14O | 114.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-49-2
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.19
InChI Key VSSAZBXXNIABDN-UHFFFAOYSA-N
Synonym cyclohexanemethanol, cyclohexylcarbinol, hexahydrobenzyl alcohol, cyclohexanecarbinol, methanol, cyclohexyl, cyclohexylmethyl alcohol, benzyl alcohol, hexahydro, usaf do-49, hydroxymethylcyclohexane, hydroxymethyl cyclohexane
PubChem CID 7507
IUPAC Name cyclohexylmethanol
SMILES C1CCC(CC1)CO

Specifications

Density 0.9100g/mL
Boiling Point 181.0°C
Flash Point 71°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4620 to 1.4650
Linear Formula C6H11CH2OH
Quantity 25 g
Beilstein 06,14
Merck Index 15,2724
Specific Gravity 0.91
Solubility Information Solubility in water: .. Other solubilities: soluble in alcohol and ether
Formula Weight 114.19
Percent Purity 99%
Chemical Name or Material Cyclohexanemethanol
Show More Show Less
EINECSNumber 202-857-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.