Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexanemethylamine, 98%

Catalog No. AAL0650106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0650106 5 g
AAL0650114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0650106 Supplier Thermo Scientific Chemicals Supplier No. L0650106
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 3218-02-8 | C7H15N | 113.20 g/mol

Cyclohexanemethylamine was used in the synthesis of Schiff?s base ligand, 4-methyl-2,6-bis(cyclohexylmethyliminomethyl)-phenol, which forms hexanuclear zinc(II) complexes. It was used during the synthesis of 1-acyl-4 cycloalkyl/arylsemicarbazides and 1-acyl-5-benzyloxy/hydroxyl carbamoylcarbazides from the nonsteroidal anti-inflammatory drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexanemethylamine was used in the synthesis of Schiff′s base ligand, 4-methyl-2,6-bis(cyclohexylmethyliminomethyl)-phenol, which forms hexanuclear zinc(II) complexes. It was used during the synthesis of 1-acyl-4 cycloalkyl/arylsemicarbazides and 1-acyl-5-benzyloxy/hydroxyl carbamoylcarbazides from the nonsteroidal anti-inflammatory drugs.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3218-02-8
Molecular Formula C7H15N
Molecular Weight (g/mol) 113.20
MDL Number MFCD00001520
InChI Key AVKNGPAMCBSNSO-UHFFFAOYSA-N
Synonym cyclohexanemethylamine, aminomethylcyclohexane, aminomethyl cyclohexane, cyclohexanemethanamine, 1-cyclohexylmethanamine, hexahydrobenzylamine, 1-cyclohexylmethylamine, 4-aminomethylcyclohexane, c-cyclohexyl-methylamine, chembl1049
PubChem CID 76688
IUPAC Name cyclohexylmethanamine
SMILES C1CCC(CC1)CN

Specifications

Melting Point -8°C
Density 0.871
Boiling Point 161°C to 163°C
Flash Point 43°C (109°F)
Refractive Index 1.462
Quantity 5 g
UN Number UN2734
Beilstein 635751
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 113.2
Percent Purity 98%
Chemical Name or Material Cyclohexanemethylamine
Show More Show Less
Hazard Category H226-H314-H335
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.
EINECSNumber 221-741-8
RTECSNumber GV6128000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.