Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexanol, ≥98.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018396 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8396 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8396 Supplier MilliporeSigma Supelco Supplier No. 441135ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 572°F
CAS 108-93-0
Vapor Pressure 0.98 mmHg (25°C)
Vapor Density 3.5 (vs. air)
Density 0.948 g/mL (at 25°C)
Flash Point 68°C
Physical Form Neat
Linear Formula C6H11 OH
MDL Number MFCD00003855
Melting Point 20°C to 22°C
Boiling Point 160°C to 161°C
Type Analytical Standard
Quantity 5 mL
Formula Weight 100.16
Percent Purity ≥98.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.