Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexanol, 98%, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
10L
Packaging:
Glass Bottle
Plastic drum
108-93-0
C6H12O
100.16
HPXRVTGHNJAIIH-UHFFFAOYSA-N
cyclohexyl alcohol,hexahydrophenol,hydrophenol,hydroxycyclohexane,hexalin,1-cyclohexanol,hydralin,adronal,naxol,adronol
7966
CHEBI:18099
cyclohexanol
C1CCC(CC1)O
108-93-0
C6H12O
100.16
HPXRVTGHNJAIIH-UHFFFAOYSA-N
cyclohexyl alcohol,hexahydrophenol,hydrophenol,hydroxycyclohexane,hexalin,1-cyclohexanol,hydralin,adronal,naxol,adronol
7966
CHEBI:18099
cyclohexanol
C1CCC(CC1)O
Specific Gravity 0.96
Packaging Glass Bottle
Infrared Spectrum Authentic
Refractive Index 1.4630 to 1.4670
Flash Point 67°C
Quantity 1L
Melting Point 23.0°C
Color Colorless
Boiling Point 161.0°C
Density 0.9600g/mL
Assay Percent Range 98%
Linear Formula C6H11OH
Beilstein 06,5
Merck Index 15,2718
Solubility Information Solubility in water: 3.6g/100mL (20°C). Other solubilities: soluble in ethanol, ethyle acetate and, aromatic hydrocarbons
Formula Weight 100.16
Physical Form Liquid After Melting
Percent Purity 98%
Chemical Name or Material Cyclohexanol
Show More Show Less