Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexanol, 98%

Catalog No. p-4510059
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC147680010 1 L Glass Bottle
AC147680025 2.5 L Glass Bottle
AC147680100 10 L Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC147680010 Supplier Thermo Scientific Chemicals Supplier No. 147680010
Only null left
Add to Cart

CAS: 108-93-0 | C6H12O | 100.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-93-0
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
InChI Key HPXRVTGHNJAIIH-UHFFFAOYSA-N
Synonym cyclohexyl alcohol, hexahydrophenol, hydrophenol, hydroxycyclohexane, hexalin, 1-cyclohexanol, hydralin, adronal, naxol, adronol
PubChem CID 7966
ChEBI CHEBI:18099
IUPAC Name cyclohexanol
SMILES C1CCC(CC1)O

Specifications

Melting Point 23.0°C
Color Colorless
Density 0.9600g/mL
Boiling Point 161.0°C
Flash Point 67°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Refractive Index 1.4630 to 1.4670
Linear Formula C6H11OH
Quantity 1 L
Beilstein 06,5
Merck Index 15,2718
Specific Gravity 0.96
Solubility Information Solubility in water: 3.6g/100mL (20°C). Other solubilities: soluble in ethanol, ethyle acetate and, aromatic hydrocarbons
Formula Weight 100.16
Percent Purity 98%
Physical Form Liquid After Melting
Chemical Name or Material Cyclohexanol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Harmful if inhaled.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protec
EINECSNumber 203-630-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.