Learn More
Medchemexpress LLC Cyclohexanone, 4-ethyl-2,6-bis(3-pyridinylmethylene)-, (2E,6E)- | 1162656-22-5 | 98.5% | 304.39 | 50 MG
SDP
Catalog No. 5000401285
Encompass_Preferred
MCB-613 is a potent Steroid receptor coactivator SRC small molecule "stimulator" (SMS) that super-stimulates SRCs' transcriptional activity. It increases SRCs' interactions with other coactivators and markedly induces ER stress coupled to the generation of reactive oxygen species (ROS). MCB-613 is a SMS that targets oncogenes and can be exploited as an anti-cancer agent by over-stimulating the SRC oncogenic program. It is for research use only and not sold to patients.
  • Potent Steroid receptor coactivator SRC small molecule "stimulator" (SMS)
  • Super-stimulates SRCs' transcriptional activity
  • Increases SRCs' interactions with other coactivators
  • Markedly induces ER stress coupled to the generation of reactive oxygen species (ROS)
  • Targets oncogenes
  • Can be exploited as an anti-cancer agent by over-stimulating the SRC oncogenic program
Catalog No. 50-004-01285 Supplier Medchemexpress LLC Supplier No. HY1962550MG
MCB-613 is a potent Steroid receptor coactivator SRC small molecule "stimulator" (SMS) that super-stimulates SRCs' transcriptional activity. It increases SRCs' interactions with other coactivators and markedly induces ER stress coupled to the generation of reactive oxygen species (ROS). MCB-613 is a SMS that targets oncogenes and can be exploited as an anti-cancer agent by over-stimulating the SRC oncogenic program. It is for research use only and not sold to patients.
  • Potent Steroid receptor coactivator SRC small molecule "stimulator" (SMS)
  • Super-stimulates SRCs' transcriptional activity
  • Increases SRCs' interactions with other coactivators
  • Markedly induces ER stress coupled to the generation of reactive oxygen species (ROS)
  • Targets oncogenes
  • Can be exploited as an anti-cancer agent by over-stimulating the SRC oncogenic program