Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexanone, 99.0%, MilliporeSigma™

Catalog No. 50947930 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-947-930 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-947-930 Supplier MilliporeSigma Supplier No. CX23357
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ACS reagent grade solvent designed for routine chemical synthesis, purification, and quality control applications.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-94-1
Molecular Weight (g/mol) 98.14

Specifications

Melting Point −31°C
pH 7 (70 g/L, H2O, 20°C)
Boiling Point 155°C (1013 hPa)
Flash Point 43°C
Assay Percent Range 99.0% min
Packaging Glass Bottle, Pre-Packed
Quantity 500 mL
Residue after Evaporation 0.05% max
Vapor Pressure 4.5 hPa (20°C)
Percent Purity ≥99.5%
Grade Not Available
Physical Form Liquid
Show More Show Less
Recommended Storage Store at room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.