Learn More
Cyclohexanone, 99+%
Description
Cyclohexanone is used in the preparation of synthetic polymer such as nylon 6,6 and nylon 6. It acts as a solvent in organic synthesis. Its derivatives are used for the synthesis of dyes, rubber chemicals, plasticizers and pharmaceuticals. Further, it is involved in the preparation of cyclohexanone oxime, which rearranges to give caprolactam in the presence of sulfuric acid catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Cyclohexanone is used in the preparation of synthetic polymer such as nylon 6,6 and nylon 6. It acts as a solvent in organic synthesis. Its derivatives are used for the synthesis of dyes, rubber chemicals, plasticizers and pharmaceuticals. Further, it is involved in the preparation of cyclohexanone oxime, which rearranges to give caprolactam in the presence of sulfuric acid catalyst.
Solubility
Miscible with diethyl ether, acetone and alcohol. Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents, red metals, lead, amines, sulfuric acid, nitric acid, strong acids, aliphatic amines, rubber, resins and alkalies.
Specifications
Specifications
| Melting Point | °C to 26°C |
| Density | 0.9478 |
| Quantity | 2500 mL |
| Flash Point | 47°C (116°F) |
| Physical Form | Liquid |
| Refractive Index | 1.4507 |
| Assay Percent Range | ≥99% |
| UN Number | UN1915 |
| Beilstein | 385735 |
| Merck Index | 14,2726 |
| Show More |
Safety and Handling
| Hazard Category | H226-H302+H312+H332-H315-H318-H335-H336 |
| Hazard Statement | GHS H Statement H226-H332 |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P314-P330-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: CYCLOHEXANONE |
| EINECSNumber | 203-631-1 |
| RTECSNumber | GW1050000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.