Learn More
Cyclohexanone, ACS, 99+%
Description
Cyclohexanone is used as an industrial solvent and paint remover. It acts as a precursor to nylon 6,6 and nylon 6 and cyclohexanone oxime, which gives caprolactam on rearrangement. Further, it is used as a chemical reaction medium, adhesives, sealants and agricultural products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | °C to 26°C |
| Color | Colorless |
| Appearance | Colorless liquid |
| Density | 0.947 g/cm3 at 20°C |
| Quantity | 1 L |
| Flash Point | 47°C (116°F) |
| Physical Form | Liquid |
| Refractive Index | 1.4507 |
| Assay Percent Range | >99% |
| Vapor Pressure | 4.5 hPa (3mm Hg) at 20°C |
| Show More |
Safety and Handling
| Hazard Category | H226-H302+H312+H332-H315-H318-H335-H336 |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P314-P330-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | DOT Class: 3; Packing Group: III |
| EINECSNumber | 203-631-1 |
| RTECSNumber | GW1050000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.