Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexanone ACS AR, Macron Fine Chemicals™

Catalog No. MK4872500 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 mL
6 x 1 L
Packaging:
Glass
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MK-4872-500 500 mL Glass
MK-4872-1 6 x 1 L Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MK-4872-500 Supplier Avantor Supplier No. 487204
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 108-94-1
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
InChI Key JHIVVAPYMSGYDF-UHFFFAOYSA-N
Synonym ketohexamethylene, pimelic ketone, sextone, cyclohexyl ketone, nadone, anone, anon, cyclohexanon, hytrol o, hexanon
PubChem CID 7967
ChEBI CHEBI:17854
IUPAC Name cyclohexanone
SMILES C1CCC(=O)CC1

Specifications

Quantity 500 mL
Packaging Glass
Formula Weight 98.14
Grade ACS
Chemical Name or Material Cyclohexanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.