Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexanone ACS, Greenfield

Catalog No. 502946098
Encompass
Change view
Click to view available options
4 x 4 L Case
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
50-294-6098 4 x 4 L Case
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-294-6098 Supplier Thermo Scientific Production Chemicals and Services Supplier No. P314000ACSCS4L
May include imposed supplier surcharges.
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 108-94-1
Molecular Weight (g/mol) 98.14

Specifications

Packaging 4x4L Case
Boiling Point 312.8 °F (156 °C)
Percent Purity 100%
Grade Reagent
Chemical Name or Material Cyclohexanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.