Learn More
Perkin Elmer US LLC Cyclohexanone - Calibration Standard, 4 g
Catalog No. 502112005
Encompass_Preferred
The PerkinElmer Combustion Standards are stable materials which are not subject to ambient influences and are recommended for the PerkinElmer 2400/2410 and Model 240 instruments.
Catalog No. 50-211-2005 Supplier Perkin Elmer US LLC Supplier No. 02401347
The PerkinElmer Combustion Standards are stable materials which are not subject to ambient influences and are recommended for the PerkinElmer 2400/2410 and Model 240 instruments.