Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexenylferrocene 95.0+%, TCI America™
SDP

Catalog No. F031225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F031225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F031225G Supplier TCI America Supplier No. F031225G

Chemical Identifiers

CAS 33183-07-2
Molecular Formula C16H18Fe
MDL Number MFCD00191435
Synonym Cyclohexenylbis(cyclopentadienyl) Iron, Ferrocenecyclohexen-1-yl

Specifications

Melting Point 65°C
Quantity 25 g
Formula Weight 266.17
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Cyclohexenylferrocene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.