Learn More
Gold Biotechnology Inc Cycloheximide 1 g
SDP
Catalog No. 5000562111
Encompass_Preferred
Cycloheximide is a protein synthesis inhibitor in eukaryotic cells It prevents ribosomal mRNA translocation during protein translation Cycloheximide has a variety of applications It can be used to study cellular processes and to determine the half-life of a protein using western blotting techniques of protein cell lysate It has been used to select for CHX-resistant strains of yeast and fungi It also displays neuroprotective properties Additionally Cycloheximide has been used to inhibit yeast growth at concentrations of 200 M It has been used to promote production of ethylene in leaves and fruit
Catalog No. 50-005-62111 Supplier Gold Biotechnology Inc Supplier No. C9301
Cycloheximide is a protein synthesis inhibitor in eukaryotic cells It prevents ribosomal mRNA translocation during protein translation Cycloheximide has a variety of applications It can be used to study cellular processes and to determine the half-life of a protein using western blotting techniques of protein cell lysate It has been used to select for CHX-resistant strains of yeast and fungi It also displays neuroprotective properties Additionally Cycloheximide has been used to inhibit yeast growth at concentrations of 200 M It has been used to promote production of ethylene in leaves and fruit