Learn More
AG Scientific Inc Cycloheximide 1 G
Catalog No. 502268256
Encompass_Preferred
Cycloheximide is an antibiotic produced by Streptomyces sp. that activates against yeasts and fungi. It inhibits protein synthesis thru DNA translation arrest in eukaryotes. Cycloheximide also inhibits chain initiation and chain elongation by acting on 60S subunits of eukaryote ribosome.
Catalog No. 50-226-8256 Supplier AG Scientific Inc Supplier No. C11891GM
Cycloheximide is an antibiotic produced by Streptomyces sp. that activates against yeasts and fungi. It inhibits protein synthesis thru DNA translation arrest in eukaryotes. Cycloheximide also inhibits chain initiation and chain elongation by acting on 60S subunits of eukaryote ribosome.