Learn More
Alkali Scientific Cycloheximide, 1 G
SDP
Catalog No. 5000660248
Encompass_Preferred
Cycloheximide is a protein synthesis inhibitor widely used in microbiology and cell biology laboratories as an antifungal agent. It functions by blocking the translocation step in eukaryotic ribosomes, effectively preventing protein synthesis in fungi and other eukaryotic organisms. This selective inhibition makes it useful in culture media where suppression of fungal growth is required while allowing bacterial growth to proceed. Cycloheximide is commonly incorporated into selective agar formulations and research protocols to control contamination by molds and yeasts. It is also used in molecular biology studies to investigate protein synthesis mechanisms and cellular responses to translational inhibition in experimental systems.
Catalog No. 50-006-60248 Supplier Alkali Scientific Supplier No. CYC001CP
Cycloheximide is a protein synthesis inhibitor widely used in microbiology and cell biology laboratories as an antifungal agent. It functions by blocking the translocation step in eukaryotic ribosomes, effectively preventing protein synthesis in fungi and other eukaryotic organisms. This selective inhibition makes it useful in culture media where suppression of fungal growth is required while allowing bacterial growth to proceed. Cycloheximide is commonly incorporated into selective agar formulations and research protocols to control contamination by molds and yeasts. It is also used in molecular biology studies to investigate protein synthesis mechanisms and cellular responses to translational inhibition in experimental systems.