Learn More
AG Scientific Inc Cycloheximide 10 G
Catalog No. 502268258
Encompass_Preferred
Cycloheximide is an antibiotic produced by Streptomyces sp. that activates against yeasts and fungi. It inhibits protein synthesis thru DNA translation arrest in eukaryotes. Cycloheximide also inhibits chain initiation and chain elongation by acting on 60S subunits of eukaryote ribosome.
Catalog No. 50-226-8258 Supplier AG Scientific Inc Supplier No. C118910GM
Cycloheximide is an antibiotic produced by Streptomyces sp. that activates against yeasts and fungi. It inhibits protein synthesis thru DNA translation arrest in eukaryotes. Cycloheximide also inhibits chain initiation and chain elongation by acting on 60S subunits of eukaryote ribosome.