Learn More
Cayman Chemical Cycloheximide, Purity: ≥95%, CAS: 66-81-9, 100 mg
SDP
Catalog No. 502955495
Encompass_Preferred
An inhibitor of protein synthesis in eukaryotes (IC50 = 5-50 μM) that functions by interfering with translational elongation; can either induce or inhibit apoptosis depending on cell type
Catalog No. 50-295-5495 Supplier Cayman Chemical Supplier No. 14126100 MG
An inhibitor of protein synthesis in eukaryotes (IC50 = 5-50 μM) that functions by interfering with translational elongation; can either induce or inhibit apoptosis depending on cell type