Learn More
Cayman Chemical Cycloheximide, 66-81-9, 100 mg
SDP
Supplier:  Cayman Chemical 14126100
Encompass_Preferred
Molecular Formula C15H23NO4, Purity ≥95%, Formula Weight 281.4, Synonyms: 4-[(2R)-2-[(1S,3S,5S)-3,5-dimethyl-2-oxocyclohexyl]-2-hydroxyethyl]-2,6-piperidinedione, Naramycin A, NSC 185; An inhibitor of protein synthesis in eukaryotes (IC50 = 5-50 M) that functions by interfering with translational elongation; can either induce or inhibit apoptosis depending on cell type.
Catalog No. NC1163264
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.