Learn More
Research Products International Corp Cycloheximide, 1 Gram
SDP
Catalog No. 50213267
Encompass_Preferred
Cycloheximide is a fungal-derived compound used particularly in molecular biology, microbiology, and cell biology. It inhibits protein synthesis in eukaryotic cells by interfering with the translocation step during protein elongation on the ribosome. It binds to the 60S ribosomal subunit and prevents the movement of peptidyl-tRNA from the A-site to the P-site, inhibiting the synthesis of new polypeptide chains. 
WARNING! This product can expose you to Cycloheximide, a chemical which is known to the State of California to cause developmental harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-213-267 Supplier Research Products International Corp Supplier No. C810401.0
Cycloheximide is a fungal-derived compound used particularly in molecular biology, microbiology, and cell biology. It inhibits protein synthesis in eukaryotic cells by interfering with the translocation step during protein elongation on the ribosome. It binds to the 60S ribosomal subunit and prevents the movement of peptidyl-tRNA from the A-site to the P-site, inhibiting the synthesis of new polypeptide chains. 
WARNING! This product can expose you to Cycloheximide, a chemical which is known to the State of California to cause developmental harm. For more information go to www.P65Warnings.ca.gov.