Learn More
Selleck Chemical LLC Cycloheximide-200mg
SDP
Supplier:  Selleck Chemical LLC S7418
Encompass_Preferred
Cycloheximide, an antifungal antibiotic, is an eukaryote protein synthesis inhibitor with IC50 of 532.5 nM and 2880 nM for protein synthesis and RNA synthesis in vivo, respectively. Cycloheximide suppresses ferroptosis and inhibits autophagy.
Catalog No. 50-238-9648