Learn More
TARGETMOL CHEMICALS INC Cycloheximide 500MG
SDP
Catalog No. 502875935
Encompass_Preferred
Also available in 200 mg and bulk. Please contact Fisher for quotes. Cycloheximide (Naramycin A) is a natural product that reversibly inhibits translation elongation, is an antimitotic compound, and is an inhibitor of DNA and protein synthesis.
Catalog No. 50-287-5935 Supplier TARGETMOL CHEMICALS INC Supplier No. T1225500MG
Also available in 200 mg and bulk. Please contact Fisher for quotes. Cycloheximide (Naramycin A) is a natural product that reversibly inhibits translation elongation, is an antimitotic compound, and is an inhibitor of DNA and protein synthesis.