Learn More
TOKU-E Cycloheximide A, EvoPure
Catalog No. NC2471751
Encompass_Preferred
Cycloheximide A, EvoPure is a highly purified (≥99.0) form of Cycloheximide A, the active compound in standard grade Cycloheximide. Cycloheximide is a glutarimide antibiotic and natural fungicide isolated from Streptomyces griseus and a protein synthesis inhibitor in eukaryotic cells.
Catalog No. NC2471751 Supplier TOKU-E Supplier No. C1231G
Cycloheximide A, EvoPure is a highly purified (≥99.0) form of Cycloheximide A, the active compound in standard grade Cycloheximide. Cycloheximide is a glutarimide antibiotic and natural fungicide isolated from Streptomyces griseus and a protein synthesis inhibitor in eukaryotic cells.