Learn More
Crescent Chemical Co Inc CYCLOHEXIMIDE CRYST. PURE
SDP
Supplier:  Crescent Chemical Co Inc SE10700.04
Encompass_Preferred
Cycloheximide cryst. pure Glutarimide antibiotic isolated from Streptomyces griseus; active against fungi and yeasts, but not against bacteria. Inhibits eukaryotic protein synthesis by blocking the translocation step in the elongation cycle (1,3,8). Blocks tra
Catalog No. 50-247-147