Learn More
Cayman Chemical Cyclohexnoyl-Coenzyme A 500ug
SDP
Catalog No. 502994674
Encompass_Preferred
An acyl-CoA that contains a cyclohexane group; formed through conversion of CHC by a succinyl-CoA transferase, and degraded by a dehydrogenase; converted to hippuric acid in submitochondrial fractions isolated from guinea pig liver
Catalog No. 50-299-4674 Supplier Cayman Chemical Supplier No. 23504500 UG
An acyl-CoA that contains a cyclohexane group; formed through conversion of CHC by a succinyl-CoA transferase, and degraded by a dehydrogenase; converted to hippuric acid in submitochondrial fractions isolated from guinea pig liver