Learn More
Cayman Chemical Cyclohexnoyl-Coenzyme A 5mg
SDP
Catalog No. 502995187
Encompass_Preferred
An acyl-CoA that contains a cyclohexane group; formed through conversion of CHC by a succinyl-CoA transferase, and degraded by a dehydrogenase; converted to hippuric acid in submitochondrial fractions isolated from guinea pig liver
Catalog No. 50-299-5187 Supplier Cayman Chemical Supplier No. 235045 MG
An acyl-CoA that contains a cyclohexane group; formed through conversion of CHC by a succinyl-CoA transferase, and degraded by a dehydrogenase; converted to hippuric acid in submitochondrial fractions isolated from guinea pig liver