Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexyl Acetate, MP Biomedicals

Catalog No. ICN211111 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN211111 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN211111 Supplier MP Biomedicals, Inc Supplier No. 0521111180

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 622-45-7
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.198
InChI Key YYLLIJHXUHJATK-UHFFFAOYSA-N
Synonym acetic acid, cyclohexyl ester, hexalin acetate, cyclohexanyl acetate, cyclohexanol, acetate, cyclohexanolazetat, adronal acetate, acetic acid cyclohexyl ester, cyclohexane acetate, cyclohexylester kyseliny octove, unii-ul0rs4h1ue
PubChem CID 12146
ChEBI CHEBI:31447
IUPAC Name cyclohexyl acetate
SMILES CC(=O)OC1CCCCC1

Specifications

Quantity 100 g
Formula Weight 142.2
Chemical Name or Material Cyclohexyl Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.