Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cyclohexyl-Alpha-Mida-Boryl Aldehyde

Catalog No. 7987541G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-875-41G 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 79-875-41G Supplier Sigma Aldrich Supplier No. 7987541G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

This boronic acid derivative is in a series of the first known stable alpha-boryl aldehyde species. Through the employment of the MIDA-protected boronic ester, this building block has been shown as a precursor to the synthesis of unnatural amino acids.Amphoteric Alpha-Boryl Aldehydes

Specifications

CAS 3087-36-3
Density 1.088 g/mL at 25°C
Boiling Point 150°C to 152°C (10 mmHg, lit.)
Molecular Formula C8H20O4Ti
Linear Formula Ti(OC2H5)4
MDL Number MFCD00009071
Quantity 50 mL
Molecular Weight (g/mol) 228.11 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.