Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexyl Butyrate 98.0+%, TCI America™
SDP

Catalog No. B075825ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B075825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B075825ML Supplier TCI America Supplier No. B075825ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1551-44-6
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.252
MDL Number MFCD00046354
InChI Key VZHUBBUZNIULNM-UHFFFAOYSA-N
Synonym cyclohexyl butyrate, butanoic acid, cyclohexyl ester, cyclohexanyl butyrate, butyric acid, cyclohexyl ester, cyclohexyl n-butyrate, unii-fhh53z3i16, fema no. 2351, ccris 6553, cyclohexanol butanoate, cyclohexyl-n-butyrate
PubChem CID 243783
IUPAC Name cyclohexyl butanoate
SMILES CCCC(=O)OC1CCCCC1

Specifications

Color Colorless
Boiling Point 208°C
Quantity 25 mL
Formula Weight 170.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclohexyl Butyrate
EINECSNumber (3)-2359
RTECSNumber ES8915000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.