Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyclohexyl ethyl ketone | 1123-86-0 | MFCD00060814 | 140.22 g/mol | C9H16O | 5 G
SDP

Catalog No. 5000301643
Encompass_Preferred

Cyclohexyl ethyl ketone (CAS 1123-86-0) is a small-molecule ketone used as a biochemical reagent in chemical synthesis and laboratory assays. It has the molecular formula C9H16O and a molecular weight of 140.22 g/mol. The material is supplied in multiple laboratory package sizes and is for research use only.

  • Small-molecule ketone reagent for synthesis and assays.
  • Molecular formula C9H16O.
  • Molecular weight 140.22 g/mol.
  • Available in multiple laboratory package sizes.
  • Recommended storage: room temperature (stable 3 years); in solvent -80°C (2 years); -20°C (1 year).
  • Intended for research use only.

Catalog No. 50-003-01643 Supplier Medchemexpress LLC Supplier No. HYW0260295G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyclohexyl ethyl ketone (CAS 1123-86-0) is a small-molecule ketone used as a biochemical reagent in chemical synthesis and laboratory assays. It has the molecular formula C9H16O and a molecular weight of 140.22 g/mol. The material is supplied in multiple laboratory package sizes and is for research use only.

  • Small-molecule ketone reagent for synthesis and assays.
  • Molecular formula C9H16O.
  • Molecular weight 140.22 g/mol.
  • Available in multiple laboratory package sizes.
  • Recommended storage: room temperature (stable 3 years); in solvent -80°C (2 years); -20°C (1 year).
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.