Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyclohexyl ethyl ketone | 1123-86-0 | MFCD00060814 | 99.3% | 140.22 g/mol | C9H16O | 250 MG
SDP

Catalog No. 5000302823
Encompass_Preferred

Cyclohexyl ethyl ketone (1-cyclohexylpropan-1-one; CAS 1123-86-0) is a small-molecule aliphatic ketone supplied for research use. It has molecular formula C9H16O, molecular weight 140.22 g/mol, and is a colorless to off-white liquid used as a biochemical reagent and synthetic intermediate in organic chemistry applications.

  • High purity (99.3%).
  • Molecular weight 140.22 g/mol.
  • Chemical formula C9H16O.
  • Liquid, colorless to off-white appearance.
  • Available in laboratory pack sizes suitable for research.

Catalog No. 50-003-02823 Supplier Medchemexpress LLC Supplier No. 250MGHYW026029
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyclohexyl ethyl ketone (1-cyclohexylpropan-1-one; CAS 1123-86-0) is a small-molecule aliphatic ketone supplied for research use. It has molecular formula C9H16O, molecular weight 140.22 g/mol, and is a colorless to off-white liquid used as a biochemical reagent and synthetic intermediate in organic chemistry applications.

  • High purity (99.3%).
  • Molecular weight 140.22 g/mol.
  • Chemical formula C9H16O.
  • Liquid, colorless to off-white appearance.
  • Available in laboratory pack sizes suitable for research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.