Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexyl Isovalerate 99.0+%, TCI America™
SDP

Catalog No. I019525ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I019525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I019525ML Supplier TCI America Supplier No. I019525ML

Chemical Identifiers

CAS 7774-44-9
Molecular Formula C11H20O2
Molecular Weight (g/mol) 184.279
MDL Number MFCD00046351
InChI Key SQPOKBBCNZIWFI-UHFFFAOYSA-N
Synonym Isovaleric Acid Cyclohexyl Ester
PubChem CID 287439
IUPAC Name cyclohexyl 3-methylbutanoate
SMILES CC(C)CC(=O)OC1CCCCC1

Specifications

Color Colorless
Boiling Point 223°C
Quantity 25 mL
Formula Weight 184.28
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclohexyl Isovalerate
EINECSNumber (3)-2359
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.