Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexyl Methyl Ketone, MP Biomedicals

Catalog No. ICN204902 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN204902 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN204902 Supplier MP Biomedicals, Inc Supplier No. 0520490205

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 823-76-7
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
InChI Key RIFKADJTWUGDOV-UHFFFAOYSA-N
Synonym 1-cyclohexylethan-1-one, cyclohexyl methyl ketone, acetylcyclohexane, ethanone, 1-cyclohexyl, cyclohexylethanone, methyl cyclohexyl ketone, 1-acetylcyclohexane, cyclohexane, acetyl, acetophenone, hexahydro, ketone, cyclohexyl methyl
PubChem CID 13207
IUPAC Name 1-cyclohexylethanone
SMILES CC(=O)C1CCCCC1

Specifications

Quantity 10 g
Formula Weight 126.2
Chemical Name or Material Cyclohexyl Methyl Ketone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.