Learn More
Medchemexpress LLC Cyclohexyl phenyl ketone | 712-50-5 | MFCD00001467 | 99.9% | 188.26 g/mol | C13H16O | 25 G
SDP
Catalog No. 5000303091
Encompass_Preferred
Cyclohexyl phenyl ketone (CAS 712-50-5) is an aromatic ketone supplied as a biochemical reagent for research and organic synthesis. Offered at high purity, it is used as an organic compound or biological material in assay development, synthetic reactions, and related life-science applications.
  • High purity suitable for research (listed as 99.93%).
  • Solid powder form, compatible with common organic solvents.
  • Useful as a building block for organic synthesis and assay reagents.
  • Available in multiple pack sizes for small-scale and bulk use.
  • Identified by CAS 712-50-5 and molecular formula C13H16O for traceability.
Catalog No. 50-003-03091 Supplier Medchemexpress LLC Supplier No. 25GHYW016067
Cyclohexyl phenyl ketone (CAS 712-50-5) is an aromatic ketone supplied as a biochemical reagent for research and organic synthesis. Offered at high purity, it is used as an organic compound or biological material in assay development, synthetic reactions, and related life-science applications.
  • High purity suitable for research (listed as 99.93%).
  • Solid powder form, compatible with common organic solvents.
  • Useful as a building block for organic synthesis and assay reagents.
  • Available in multiple pack sizes for small-scale and bulk use.
  • Identified by CAS 712-50-5 and molecular formula C13H16O for traceability.