Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyclohexyl phenyl ketone | 712-50-5 | MFCD00001467 | 99.9% | 188.27 g/mol | C13H16O | 100 G
SDP

Supplier:  Medchemexpress LLC HYW016067100G

Encompass_Preferred

Cyclohexyl phenyl ketone (CAS 712-50-5) is an organic ketone reagent used in life-science research for biochemical assays, synthetic transformations, and analytical studies. It is supplied as a solid with characterized physicochemical properties and high assay purity suitable for laboratory use.

  • High purity: 99.9% by assay.
  • Molecular formula: C13H16O; molecular weight: 188.27 g/mol.
  • Solid form suitable for handling and storage.
  • Recommended storage: powder at -20°C (long term) or 4°C (short term); in solvent at -80°C.
  • Suitable for analytical, synthetic, and assay applications.

Catalog No. 50-003-02883


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.