Learn More
Medchemexpress LLC Cyclohexyl phenyl ketone | 712-50-5 | MFCD00001467 | 99.9% | 188.27 g/mol | C13H16O | 5 G
SDP
Catalog No. 5000302144
Encompass_Preferred
Cyclohexyl phenyl ketone (CAS 712-50-5) is an aromatic ketone (C13H16O, MW 188.27 g/mol) used as a biochemical reagent and intermediate in organic synthesis. Supplied at high purity for research applications, it is commonly used as a reference standard, in laboratory assays, and as a building block for chemical synthesis.
  • High purity for analytical and synthetic applications.
  • Well-defined chemical identity (C13H16O; MW 188.27 g/mol).
  • Suitable as a reference standard and synthetic intermediate.
  • Available in multiple pack sizes to support small- and bulk-scale work.
  • Provided with safety and handling documentation (SDS) for laboratory use.
Catalog No. 50-003-02144 Supplier Medchemexpress LLC Supplier No. HYW0160675G
Cyclohexyl phenyl ketone (CAS 712-50-5) is an aromatic ketone (C13H16O, MW 188.27 g/mol) used as a biochemical reagent and intermediate in organic synthesis. Supplied at high purity for research applications, it is commonly used as a reference standard, in laboratory assays, and as a building block for chemical synthesis.
  • High purity for analytical and synthetic applications.
  • Well-defined chemical identity (C13H16O; MW 188.27 g/mol).
  • Suitable as a reference standard and synthetic intermediate.
  • Available in multiple pack sizes to support small- and bulk-scale work.
  • Provided with safety and handling documentation (SDS) for laboratory use.