Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexyl Phenyl Ketone, MP Biomedicals

Catalog No. ICN210694 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN210694 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN210694 Supplier MP Biomedicals, Inc Supplier No. 0521069405

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 712-50-5
Molecular Formula C13H16O
Molecular Weight (g/mol) 188.27
InChI Key BMFYCFSWWDXEPB-UHFFFAOYSA-N
Synonym cyclohexyl phenyl ketone, cyclohexyl phenyl methanone, benzoylcyclohexane, cyclohexylphenylketone, benzoyl cyclohexane, phenyl cyclohexyl ketone, methanone, cyclohexylphenyl, cyclohexyl-phenyl-methanone, usaf kf-3, cyclohexylphenyl ketone
PubChem CID 12837
IUPAC Name cyclohexyl(phenyl)methanone
SMILES C1CCC(CC1)C(=O)C2=CC=CC=C2

Specifications

Quantity 10 g
Formula Weight 188.27
Chemical Name or Material Cyclohexyl Phenyl Ketone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.