Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexylacetic acid, 98+%

Catalog No. AC111271000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC111271000 100 g Glass bottle
AC111270050 5 g Glass bottle
AC111270250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC111271000 Supplier Thermo Scientific Chemicals Supplier No. 111271000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 5292-21-7 | C8H14O2 | 142.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5292-21-7
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00001518
InChI Key LJOODBDWMQKMFB-UHFFFAOYSA-N
Synonym cyclohexylacetic acid, cyclohexaneacetic acid, cyclohexylethanoic acid, hexahydrophenylacetic acid, cyclohexyl acetic acid, unii-v43k3to0hn, fema no. 2347, cyclohexane-acetic acid, v43k3to0hn, cyclo-hexylacetic acid
PubChem CID 21363
ChEBI CHEBI:37277
IUPAC Name 2-cyclohexylacetic acid
SMILES OC(=O)CC1CCCCC1

Specifications

Melting Point 28.0°C to 32.0°C
Color White to Yellow
Density 1.0070g/mL
Boiling Point 242.0°C to 244.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula C6H11CH2CO2H
Quantity 100 g
Beilstein 09, II, 9
Specific Gravity 1.007
Formula Weight 142.2
Percent Purity 98+%
Physical Form Low Melting Solid
Chemical Name or Material Cyclohexylacetic acid, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 226-132-0
RTECSNumber GU6490000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.