Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexylacetic acid, 98+%, Thermo Scientific™

Catalog No. AC111275000
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC111275000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC111275000 Supplier Thermo Scientific Chemicals Supplier No. 111275000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5292-21-7
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00001518
InChI Key LJOODBDWMQKMFB-UHFFFAOYSA-N
Synonym cyclohexylacetic acid, cyclohexaneacetic acid, cyclohexylethanoic acid, hexahydrophenylacetic acid, cyclohexyl acetic acid, unii-v43k3to0hn, fema no. 2347, cyclohexane-acetic acid, v43k3to0hn, cyclo-hexylacetic acid
PubChem CID 21363
ChEBI CHEBI:37277
IUPAC Name 2-cyclohexylacetic acid
SMILES OC(=O)CC1CCCCC1

Specifications

Melting Point 28.0°C to 32.0°C
Density 1.0070g/mL
Boiling Point 242.0°C to 244.0°C
Flash Point >110°C
Linear Formula C6H11CH2CO2H
Quantity 500 g
Specific Gravity 1.007
Formula Weight 142.2
Chemical Name or Material Cyclohexylacetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.