Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexylacetone, 95%

Catalog No. AAA1274806
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1274806 5 g
AAA1274822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1274806 Supplier Thermo Scientific Chemicals Supplier No. A1274806
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 103-78-6 | C9H16O | 140.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-78-6
Molecular Formula C9H16O
Molecular Weight (g/mol) 140.23
MDL Number MFCD00019396
InChI Key UHBZEAPZATVYKV-UHFFFAOYSA-N
Synonym cyclohexylacetone, 1-cyclohexylacetone, 2-propanone, 1-cyclohexyl, acetonylcyclohexane, cyclohexyl-2-propanone, 1-cyclohexyl-2-propanone, cyclohexylaceton, acmc-20alcn, 1-cyclohexylacetone #, 1-cyclohexyl-propan-2-one
PubChem CID 66899
IUPAC Name 1-cyclohexylpropan-2-one
SMILES CC(=O)CC1CCCCC1

Specifications

Density 0.905
Boiling Point 198°C to 200°C
Refractive Index 1.4525
Quantity 5 g
Beilstein 1209723
Formula Weight 140.23
Percent Purity 97%
Chemical Name or Material Cyclohexylacetone
EINECSNumber 203-143-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.