Learn More
Strem, An Ascensus Company CAS# 436865-11-1. 25g. Cyclohexyldi-t-butylphosphine, 98% (10wt% in hexanes).
SDP
Supplier:  Strem, An Ascensus Company 15101125G
Encompass
CAS# 436865-11-1. 25g. Cyclohexyldi-t-butylphosphine, 98% (10wt% in hexanes). . Molecular Weight: 228.36. Molecular Formula: C14H29P. Color/form: colorless liq. Strem# 15-1011. http://www.strem.com/catalog/v/15-1011/
Catalog No. NC1889547