Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cyclohexyldichlorophosphine

Catalog No. 6982451G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-824-51G 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 69-824-51G Supplier Sigma Aldrich Supplier No. 6982451G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Cyclohexyldichlorophosphine

Specifications

CAS 68166-83-6
Melting Point >300°C
Molecular Formula C58H48Cl2N2P2Ru
Refractive Index n20/D 1.528
Linear Formula C58H48Cl2N2P2Ru
MDL Number MFCD02684565
Quantity 100 mg
Synonym Cyclohexylphosphonous dichloride; Cyclohexylphosphorus dichloride; Dichlorocyclohexylphosphin
Molecular Weight (g/mol) 1006.94 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.