Learn More
Medchemexpress LLC Cyclohexyldiphenylphosphine | 6372-42-5 | 99.99% | 268.34 | 1 KG
SDP
Catalog No. 5000454399
Encompass_Preferred
Cyclohexyldiphenylphosphine is a chemical compound frequently employed as a ligand in coordination chemistry and organometallic synthesis. It acts as a stabilizing agent, facilitating various catalytic reactions. This compound also serves as a ligand for certain nickel(II) complexes and has demonstrated anti-cancer properties against SNO cancer cells when complexed with silver.
  • Commonly used as a ligand in coordination chemistry.
  • Utilized in organometallic synthesis.
  • Facilitates various catalytic reactions.
  • Acts as a stabilizing agent.
  • Serves as a ligand for nickel(II) complexes.
  • Exhibits anti-cancer properties when complexed with silver.
Catalog No. 50-004-54399 Supplier Medchemexpress LLC Supplier No. HYW0133271KG
Cyclohexyldiphenylphosphine is a chemical compound frequently employed as a ligand in coordination chemistry and organometallic synthesis. It acts as a stabilizing agent, facilitating various catalytic reactions. This compound also serves as a ligand for certain nickel(II) complexes and has demonstrated anti-cancer properties against SNO cancer cells when complexed with silver.
  • Commonly used as a ligand in coordination chemistry.
  • Utilized in organometallic synthesis.
  • Facilitates various catalytic reactions.
  • Acts as a stabilizing agent.
  • Serves as a ligand for nickel(II) complexes.
  • Exhibits anti-cancer properties when complexed with silver.