Learn More
Medchemexpress LLC Cyclohexyldiphenylphosphine | 6372-42-5 | 99.99% | 268.34 | 25 G
SDP
Catalog No. 5000366701
Encompass_Preferred
Cyclohexyldiphenylphosphine is a chemical compound commonly used as a ligand in coordination chemistry and organometallic synthesis. It facilitates various catalytic reactions by acting as a stabilizing agent and is a ligand for some nickel(II) complexes. This compound also exhibits anti-cancer properties against SNO cancer cells when complexed with silver.
  • Used as a ligand in coordination chemistry and organometallic synthesis.
  • Facilitates various catalytic reactions through acting as a stabilizing agent.
  • Ligand for some nickel(II) complexes.
  • Exhibits anti-cancer properties against SNO cancer cells when formed complex with silver.
Catalog No. 50-003-66701 Supplier Medchemexpress LLC Supplier No. 25GHYW013327
Cyclohexyldiphenylphosphine is a chemical compound commonly used as a ligand in coordination chemistry and organometallic synthesis. It facilitates various catalytic reactions by acting as a stabilizing agent and is a ligand for some nickel(II) complexes. This compound also exhibits anti-cancer properties against SNO cancer cells when complexed with silver.
  • Used as a ligand in coordination chemistry and organometallic synthesis.
  • Facilitates various catalytic reactions through acting as a stabilizing agent.
  • Ligand for some nickel(II) complexes.
  • Exhibits anti-cancer properties against SNO cancer cells when formed complex with silver.