Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexylhydrazine hydrochloride, 98%

Catalog No. AC430020250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC430020250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC430020250 Supplier Thermo Scientific Chemicals Supplier No. 430020250

missing translation for 'validMainframeMsg'

CAS: 24214-73-1 | C6H14N2·ClH | 150.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24214-73-1
Molecular Formula C6H14N2·ClH
Molecular Weight (g/mol) 150.65
MDL Number MFCD00060160
InChI Key JZRHODNPRNTXKO-UHFFFAOYSA-N
Synonym cyclohexylhydrazine hydrochloride, 1-cyclohexylhydrazine hydrochloride, cyclohexylhydrazinehydrochloride, hydrazine, cyclohexyl-, hydrochloride, 1-cyclohexylhydrazinehydrochloride, cyclohexyl hydrazine hydrochloride, hydrazine, cyclohexyl-, monohydrochloride, cyclohexyl-hydrazine hydrochloride, cyclohexyl-hydrazin;, cyclohexyl-hydrazine;
PubChem CID 207836
IUPAC Name cyclohexylhydrazine;hydrochloride
SMILES C1CCC(CC1)NN.Cl

Specifications

Melting Point 110.0°C to 113.0°C
Color White
Elemental Analysis 46.64 to 49.03% C, 18.13 to 19.07% N
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Formula Weight 150.65
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Cyclohexylhydrazine hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.