Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexylhydrazine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. C05005G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C05005G 5 g
C050025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C05005G Supplier TCI America Supplier No. C05005G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 24214-73-1
Molecular Formula C6H15ClN2
Molecular Weight (g/mol) 150.65
MDL Number MFCD00060160
InChI Key JZRHODNPRNTXKO-UHFFFAOYSA-N
Synonym cyclohexylhydrazine hydrochloride, 1-cyclohexylhydrazine hydrochloride, cyclohexylhydrazinehydrochloride, hydrazine, cyclohexyl-, hydrochloride, 1-cyclohexylhydrazinehydrochloride, cyclohexyl hydrazine hydrochloride, hydrazine, cyclohexyl-, monohydrochloride, cyclohexyl-hydrazine hydrochloride, cyclohexyl-hydrazin;, cyclohexyl-hydrazine;
PubChem CID 207836
IUPAC Name cyclohexylhydrazine;hydrochloride
SMILES C1CCC(CC1)NN.Cl

Specifications

Melting Point 112°C
Color White-Yellow
Quantity 5 g
Formula Weight 150.65
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Cyclohexylhydrazine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.