Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cyclohexylmagnesium Chloride Solution

Catalog No. 774677100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-467-7100ML 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 77-467-7100ML Supplier Sigma Aldrich Supplier No. 774677100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product, 1.0 M in 2-methyltetrahydrofuran aligns with Safer Solvents and Auxiliaries, Use of Renewable Feedstocks and Inherently Safer Chemistry for Accident Prevention. Click here for more information.Nucleophilic reagent is commonly used in substitution and addition reactions.Development of GSK's reagent guides - embedding sustainability into reagent selection.Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 144490-03-9
Melting Point 80°C to 83°C
Molecular Formula C13H18O9
Linear Formula C13H18O9
MDL Number MFCD07698606
Quantity 10 g
Synonym 1,2,3,5-Tetra-O-acetyl-beta-L-ribofuranose
Molecular Weight (g/mol) 318.28 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.