Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexylmethyl Acetate 98.0+%, TCI America™
SDP

Catalog No. C24435G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C24435G 5 g
C244325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C24435G Supplier TCI America Supplier No. C24435G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 937-55-3
Molecular Formula C9H16O2
Molecular Weight (g/mol) 156.23
MDL Number MFCD07780584
InChI Key FXKHUBNHBYCRNH-UHFFFAOYSA-N
Synonym Acetic Acid Cyclohexylmethyl Ester
PubChem CID 244762
IUPAC Name cyclohexylmethyl acetate
SMILES CC(=O)OCC1CCCCC1

Specifications

Color Yellow
Boiling Point 183°C
Quantity 5 g
Formula Weight 156.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclohexylmethyl Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.