Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (Cyclohexylmethyl)Magnesium Bromide Solution

Catalog No. 68795250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68-795-250ML 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 68-795-250ML Supplier Sigma Aldrich Supplier No. 68795250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant involved in:Isomerization polymerization of alkenylcyclohexanesIntramolecular rearrangements of vinylidenecyclopropanesCycloisomerization of cyclic and acyclic enynesAdditionally reactant involved in synthesis of biologically active molecules including:Dipeptidyl boronic acid proteasome inhibitorsSubstituted 1,3-dihydroindole-2-ones with antitumor effectsSulfur-free transition state nucleoside analog inhibitors of methylthioadenosine nucleosidase and phosphorylase

Specifications

CAS 137037-21-9
Melting Point 117°C to 118°C; 117°C to 118°C
Molecular Formula C8H16N2 -+ C4H6O6 -+ 3H2O
Linear Formula C8H16N2 -+ C4H6O6 -+ 3H2O
MDL Number MFCD11656040
Quantity 5 g
Synonym (2R,2'R)-2,2'-Bipyrrolidine (2R,3R)-2,3-dihydroxybutanedioate
Molecular Weight (g/mol) 344.36 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.