Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexylthiourea 98.0+%, TCI America™
SDP

Catalog No. C2966200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2966200MG 200 mg
C29661G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2966200MG Supplier TCI America Supplier No. C2966200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5055-72-1
Molecular Formula C7H14N2S
Molecular Weight (g/mol) 158.26
MDL Number MFCD00060341
InChI Key LEEHHPPLIOFGSC-UHFFFAOYSA-N
PubChem CID 1471603
IUPAC Name cyclohexylthiourea
SMILES NC(=S)NC1CCCCC1

Specifications

Melting Point 164°C
Color White-Yellow
Quantity 200 mg
Formula Weight 158.26
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Cyclohexylthiourea
RTECSNumber YS7700000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.